Filter tips, GentleFit™ Dual-Shot, Axygen®
Leverandør: Corning
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
613-2309EA 1390 DKK
AXYGTLF-1000-R-S AXYGTLF-10-R-S AXYGTLF-200-R-S
Filter tips, GentleFit™ Dual-Shot, Axygen®
These filter barrier tips are ideal for DNA amplification and microbiological applications where trace contamination can completely invalidate results.
  • PP body and TPE top, sterile
  • RNase, DNase, and endotoxin free
  • Tips create a leak proof seal: Minimal force required to load and eject tip
  • Reduced wear on pipettors – giving them a longer life
  • Ergonomic: Soft tip allows for less pressure to create an optimal seal
  • Top collar color-coded by volume