Welding face shields, Blocus
Leverandør: Bollé Safety
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Blocus
111-0974EA 0 DKK
111-0974
Welding face shields, Blocus
Ansigtsskærm
Zytel head helmet with face shield for electric arc welding applications.
  • With fixed or flip-up window
Weight: 435 g
Certificeringer: EN 175
Bestillingsoplysninger: Mask is supplied empty without any filter or headgear.