Industrial gloves, Picolon Confort
Leverandør: Ansell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
112-2830EA 392 DKK
112-2830 112-2829
Industrial gloves, Picolon Confort
Handsker
Seamless knitted glove with latex knit wrist and dot coating on palm and fingers. The textile liner is made of high elasticity polyamid thread.
  • Lightweight and lint-free
  • Excellent dexterity, good resistance to abrasion and small blade cut hazards
  • Blue PVC dots on the palm provide elevated levels of grip
  • Glove designed to protect handled objects from fingerprints
Ideal for the handling and assembly of slightly sharp and slippery objects.
Certificeringer: EN 388:2016 (2142A); Cat. II