Earmuffs, Leightning® L1
Leverandør: Howard Leight by Honeywell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Leightning®
111-1411EA 176 DKK
111-1411
Earmuffs, Leightning® L1
Høreværn Støjblokerende høreværn
Robust and durable earmuffs for rough industrial environments.
  • Low profile cups
  • Air Flow Control™ Technology provides better, consistent attenuation, especially at low frequencies
  • Snap in ear cushions makes cushion replacement quick and easy
  • Padded foam headband delivers long wearing comfort and reduces the pressure on the head
  • Telescopic height adjustment remains fixed during use
The earmuffs work especially well in extreme temperatures/humidity and under aggressive chemical influences.
SNR: 28 dB (helmet-mounted)
SNR: 29 dB (neckband)
SNR: 30 dB (headband)
Weight: 201 g (headband); 192 g (helmet-mounted); 171 g (neckband)
Certificeringer: EN 352-1