Safety spectacles, Universal
Leverandør: Bollé Safety
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Universal
111-1026EA 116 DKK
111-1026
Safety spectacles, Universal
Safety glasses with extremely comfortable fit thanks to the flexible, lightweight bridge and the pivoting temples.
  • Anti-scratch, anti-fog coatings
  • Upper and lower protection
  • Non slip B-Flex nose bridge
  • Pivoting, co-injected straight temples
Weight: 26 g
Certificeringer: EN 160, EN 170