VWR® NEXT GEL® Acrylamide Solutions
Leverandør: Avantor
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VWR®
M256-500MLEA 1860 DKK
M256-500ML M258-500ML M255-500ML M257-100ML M255-100ML M257-500ML
VWR® NEXT GEL® Acrylamide Solutions
Elektroforese geler Agarose og Akrylamid
NEXT GEL® is a ready to pour pre-mixed solution of acrylamide, bis-acrylamide, gel buffer, and SDS that enables superior, ultra-fine resolution of protein bands. The unique acrylamide matrix slows the migration of proteins, eliminating the need for a stacking gel. This not only saves time, but also allows the proteins to run across a longer gel surface resulting in increased resolution. The proprietary chemistry has gradient-like properties, which enables the separation of small peptides and high molecular weight proteins in the same gel.
  • Ready to pour SDS polyacrylamide solutions - just add APS and TEMED
  • Faster gel casting with no stacking gel required
  • Broad range of separation - 3,5 to 212 kDa on the same gel
  • High resolution of protein bands
Leveringsinformation: Supplied with 20X running buffer.