Safety shoes, lace-up, Jalcordee X2
Leverandør: Jallatte
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
JALCJ0339/43EA 1880 DKK
JALCJ0339/43 JALCJ0339/44 JALCJ0339/40 JALCJ0339/41 JALCJ0339/45 JALCJ0339/47 JALCJ0339/42 JALCJ0339/39 JALCJ0339/46
Safety shoes, lace-up, Jalcordee X2
Metal-free safety shoes made from perforated suede leather, 3D lining and a Softane™/Néotril sole (resistant against acids, hydrocarbons and direct contact with heat sources up to 300 °C); with padded tongue.
  • Lining with three dimensional foam structures for excellent ventilation
  • Confort-Tech integrated insole: Anatomical, antibacterial, shock absorber heel
  • Lightane™ sole made from Softane™/Néotril with anti-torision system to optimise the foot stability
  • Non magnetic, lightweight composite toecap (200 J)
  • Non magnetic, lightweight and flexible FleXtane™ HP midsole (100% composite)
Colour: Grey
Certificeringer: EN ISO 20345:2007; S1P, HRO, SRC