Temperature loggers, TomKey®
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
TomKey®
AESLMD5301EA 1050 DKK
AESLMD5301 AESLMD5300
Temperature loggers, TomKey®
Dataloggere
The TomKey® temperature loggers are specifically designed for temperature monitoring. They are suitable for use in thermo-sensitive products, cold chain contro and transport condition records.
  • Multi-function LCD with minimum/maximum alarms display
  • USB 2.0 interface for quick and easy communication with the software
  • Logging activation is either programmable or with the included magnet key
Leveringsinformation: Supplied with free registration and data processing software, and a calibration certificate.