Air and nitrogen generators, ANG range
Leverandør: Peak Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
PEAK10-1611EA 0 DKK
PEAK10-1611 PEAK10-1710 PEAK10-1510 PEAK10-1660
Air and nitrogen generators, ANG range
Gasgeneratorer Nitrogengeneratorer
The ANG range is designed specifically for use with GC applications.
  • Units are available with or without internal air compressors
Purity: 99,9995%
Outlet port: 1×¼" BSP female
Pressure dewpoint: –70 °C
Max outlet pressure: 5,5 bar
* nitrogen; ** dry air