Safety shoes, lace-up, Jalarthur
Leverandør: Jallatte
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
JALCJMI04/45EA 1240 DKK
JALCJMI04/45 JALCJMI04/47 JALCJMI04/39 JALCJMI04/46 JALCJMI04/42 JALCJMI04/41 JALCJMI04/40 JALCJMI04/44 JALCJMI04/43
Safety shoes, lace-up, Jalarthur
Metal-free safety shoes made from full grain leather upper, leather lining and Softane™ dual-density PU sole (resistant to abrasion, hydrocarbons, oils and grease).
  • Anatomically shaped Confort-Tech insole; anti-static and antibacterial, with shock absorber heel
  • Lightweight, non magnetic, perforated composite toecap (200 J)
  • Non magnetic, lightweight and flexible FleXtane™ HP midsole (100% composite)
  • Softane™ dual-density PU provides excellent absorption of heel shocks and vibrations
Colour: Black
Certificeringer: EN ISO 20345:2004; S1P, SRC