4'-Aminobenzo-15-crown-5 (4'-aminobenzo-1,4,7,10,13-pentaoxacyclopentadecane) ≥97.0% (by GC, titration analysis)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
TCIAA1404-1GEA
1310
DKK
TCIAA1404-1G
4'-Aminobenzo-15-crown-5 (4'-aminobenzo-1,4,7,10,13-pentaoxacyclopentadecane) ≥97.0% (by GC, titration analysis)
4'-Aminobenzo-15-crown-5 (4'-aminobenzo-1,4,7,10,13-pentaoxacyclopentadecane)
Formel:
C₁₄H₂₁NO₅ Molekylvægt: 283,3202 g/mol |
MDL Number:
MFCD00068686 CAS-nummer: 60835-71-4 |
![]() |
Specifikationer Testresultater
Appearance | White to Gray to Brown powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Melting point | 76.0 to 80.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...