m-Phenylenediamine sulphate ≥98.0% (by titrimetric analysis)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

TCIAP0174-25GEA 567 DKK
TCIAP0174-25G
m-Phenylenediamine sulphate ≥98.0% (by titrimetric analysis)
m-Phenylenediamine sulphate
Formel: C₆H₁₀N₂O₄S
MDL Number: MFCD00060223
CAS-nummer: 541-70-8
EINECS: 208-791-6
UN nummer: 2811
ADR: 6.1,III
https://dk.vwr.com/stibo/web/std.lang.all/09/08/28990908.jpg

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Purple powder to crystal
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR