3,5-Dichloro-4-pyridone ≥98.0% (by titrimetric analysis)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

TCIAD2103-25GEA 1030 DKK
TCIAD2103-25G
3,5-Dichloro-4-pyridone ≥98.0% (by titrimetric analysis)
3,5-Dichloro-4-pyridone
Formel: C₅H₃Cl₂NO
Molekylvægt: 163,9894 g/mol
MDL Number: MFCD00051673
CAS-nummer: 17228-70-5
EINECS: 241-267-5
https://dk.vwr.com/stibo/web/std.lang.all/88/84/28978884.jpg

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR