Rhodizonic acid disodium salt ≥90.0% (by titrimetric analysis)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

TCIAR0019-25GEA 2910 DKK
TCIAR0019-25G
Rhodizonic acid disodium salt ≥90.0% (by titrimetric analysis)
Rhodizonic acid disodium salt
Formel: C₆Na₂O₆
Molekylvægt: 214,04 g/mol
MDL Number: MFCD00001650
CAS-nummer: 523-21-7
EINECS: 208-340-3
https://dk.vwr.com/stibo/web/std.lang.all/37/47/28993747.jpg

Bestil nu

Specifikationer Testresultater

Appearance Yellow to Brown to Dark green powder to crystal
Purity (Nonaqueous Titration) min. 90.0 %
Drying loss max. 10.0 %
Sensitiveness to pass test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR