POPSO (β,β'-dihydroxypiperazine-1,4-dipropanesulphonic acid) good's buffer component for biological research

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

TCIAP1131-25GEA 165 DKK
TCIAP1131-25G
POPSO (β,β'-dihydroxypiperazine-1,4-dipropanesulphonic acid) good's buffer component for biological research
POPSO (β,β'-dihydroxypiperazine-1,4-dipropanesulphonic acid)
Formel: C₁₀H₂₂N₂O₈S₂
Molekylvægt: 362,4203 g/mol
MDL Number: MFCD00038350
CAS-nummer: 68189-43-5
EINECS: 269-199-1
https://dk.vwr.com/stibo/web/std.lang.all/17/60/28991760.jpg

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 % (calcd.on dried substance)
Absorbance max. 0.15 (1mol/L、1mol/L NaOH、260nm)
Solubility in 1mol/L NaOH almost transparency
Drying loss 8.0 to 10.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR