1,2-Bis(di(pentafluorophenyl)phosphino)ethane ≥97.0% (by HPLC)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
TCIAB3428-1GEA
796
DKK
TCIAB3428-1G
1,2-Bis(di(pentafluorophenyl)phosphino)ethane ≥97.0% (by HPLC)
1,2-Bis(di(pentafluorophenyl)phosphino)ethane
Formel:
C₂₆H₄F₂₀P₂ Molekylvægt: 758,226 g/mol |
MDL Number:
MFCD00058841 CAS-nummer: 76858-94-1 ADR: 6.1,III |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Melting point | 193.0 to 197.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...