Dibenzo-15-crown-5-(dibenzo-1,4,7,10,13-pentaoxacyclopentadecane) ≥95.0% (by GC)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

TCIAD2877-1GEA 1190 DKK
TCIAD2877-1G
Dibenzo-15-crown-5-(dibenzo-1,4,7,10,13-pentaoxacyclopentadecane) ≥95.0% (by GC)
Dibenzo-15-crown-5-(dibenzo-1,4,7,10,13-pentaoxacyclopentadecane)
Formel: C₁₈H₂₀O₅
Molekylvægt: 316,35 g/mol
Smeltepunkt: 115°C
MDL Number: MFCD00055949
CAS-nummer: 14262-60-3
https://dk.vwr.com/stibo/web/std.lang.all/00/33/28980033.jpg

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light red powder to crystal
Purity (GC) min. 95.0 %
Melting point 111.0 to 116.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR