1,4-Di(chlormethyl)-2,3,5,6-tetramethylbenzen ≥98.0% (by GC)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

TCIAB1783-5GEA 796 DKK
TCIAB1783-5G
1,4-Di(chlormethyl)-2,3,5,6-tetramethylbenzen ≥98.0% (by GC)
1,4-Di(chlormethyl)-2,3,5,6-tetramethylbenzen
Formel: C₁₂H₁₆Cl₂
Molekylvægt: 231,16 g/mol
MDL Number: MFCD00018883
CAS-nummer: 3022-16-0
EINECS: 221-173-0
UN nummer: 3261
ADR: 8,III
https://dk.vwr.com/stibo/web/std.lang.all/35/44/28933544.jpg

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 195.0 to 199.0 deg-C
Solubility in Toluene almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR