4,4'-Bis(dimethylamino)thiobenzophenon ≥97.0% (by titrimetric analysis) til spektrofotometri
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
TCIAA5201-1GEA
214
DKK
TCIAA5201-1G
4,4'-Bis(dimethylamino)thiobenzophenon ≥97.0% (by titrimetric analysis) til spektrofotometri
4,4'-Bis(dimethylamino)thiobenzophenon
Sensitive spectrophotometric reagent for Au, Ag, Hg and Pd, use for the determination of residual chlorine.
Formel:
C₁₇H₂₀N₂S Molekylvægt: 284,43 g/mol |
MDL Number:
MFCD00040477 CAS-nummer: 1226-46-6 EINECS: 214-962-6 |
![]() |
Specifikationer Testresultater
Appearance | Red to Dark red to Dark purple powder to crystal |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...