Pentafluoriodbenzen ≥99.0% (by GC) stabilised
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Stabilization:
Stabilised with copper chip
TCIAP1188-5GEA
238
DKK
TCIAP1188-5G
TCIAP1188-25G
Pentafluoriodbenzen ≥99.0% (by GC) stabilised
Pentafluoriodbenzen
Formel:
C₆F₅I Molekylvægt: 293,96 g/mol Kogepunkt: 162°C Smeltepunkt: –29 °C |
MDL Number:
MFCD00001032 CAS-nummer: 827-15-6 EINECS: 212-565-2 |
![]() |
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...