4-Octyloxybenzoesyre ≥98.0% (by GC, titration analysis)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
TCIAO0117-25GEA
690
DKK
TCIAO0117-25G
4-Octyloxybenzoesyre ≥98.0% (by GC, titration analysis)
4-Octyloxybenzoesyre
Formel:
C₁₅H₂₂O₃ Molekylvægt: 250,34 g/mol Smeltepunkt: 100 °C |
MDL Number:
MFCD00013993 CAS-nummer: 2493-84-7 |
![]() |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in hot Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...