Vanadyl (IV)acetylacetonat ≥95.0% (by titrimetric analysis)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

TCIAV0016-25GEA 238 DKK
TCIAV0016-25G
Vanadyl (IV)acetylacetonat ≥95.0% (by titrimetric analysis)
Vanadyl (IV)acetylacetonat
Formel: C₁₀H₁₄O₅V
Molekylvægt: 265,16 g/mol
MDL Number: MFCD00000032
CAS-nummer: 3153-26-2
EINECS: 221-590-8
ADR: 6.1,III
https://dk.vwr.com/stibo/web/std.lang.all/72/68/28997268.jpg

Bestil nu

Specifikationer Testresultater

Appearance Blue to Dark blue powder to crystal
Purity (Chelometric Titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR