Perfluorheptan (isomerblanding) ≥80.0% (by GC)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
TCIAP0851-10GEA
181
DKK
TCIAP0851-10G
Perfluorheptan (isomerblanding) ≥80.0% (by GC)
Perfluorheptan (isomerblanding)
Formel:
C₇F₁₆ Molekylvægt: 388,05 g/mol Kogepunkt: 82°C |
MDL Number:
MFCD00040339 CAS-nummer: 335-57-9 EINECS: 206-392-1 |
![]() |
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 80.0% (total of isomer) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...