Amantadinsulfat ≥98.0% (by titrimetric analysis)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

TCIAA1319-25GEA 296 DKK
TCIAA1319-25G
Amantadinsulfat ≥98.0% (by titrimetric analysis)
Amantadinsulfat
Formel: C₂₀H₃₆NO₄S
Molekylvægt: 386,58 g/mol
Smeltepunkt: 299…301 °C
MDL Number: MFCD00077251
CAS-nummer: 31377-23-8
EINECS: 250-604-5
https://dk.vwr.com/stibo/web/std.lang.all/92/25/28929225.jpg

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR