(3-Brompropyl)triphenylphosphoniumbromid ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

TCIAB1208-25GEA 321 DKK
TCIAB1208-25G
(3-Brompropyl)triphenylphosphoniumbromid ≥98.0% (by HPLC, titration analysis)
(3-Bromopropyl)triphenylphosphonium bromide
Formel: C₂₁H₂₁Br₂P
Molekylvægt: 464,18 g/mol
Smeltepunkt: 225…229 °C
MDL Number: MFCD00011866
CAS-nummer: 3607-17-8
EINECS: 222-770-9
https://dk.vwr.com/stibo/web/std.lang.all/30/44/28933044.jpg

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 221.0 to 228.0 deg-C
Solubility in hot Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR