3-Chlorstyren ≥98.0% (by GC) stabilised
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Stabilization:
stabiliseret med 4-tert-Butylpyrocatechol
TCIAC0289-10MLEA
1830
DKK
TCIAC0289-10ML
3-Chlorstyren ≥98.0% (by GC) stabilised
3-Chlorstyren
Formel:
C₈H₇Cl Molekylvægt: 138,6 g/mol Kogepunkt: 87°C Smeltepunkt: –26 °C Flammepunkt: 63°C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00000598 CAS-nummer: 2039-85-2 EINECS: 218-024-7 |
![]() |
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...