(±)-δ-Dodecalacton ≥98.0% (by GC)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
TCIAD1414-25MLEA
214
DKK
TCIAD1414-25ML
(±)-δ-Dodecalacton ≥98.0% (by GC)
(±)-δ-Dodecalacton
Formel:
C₁₂H₂₂O₂ Molekylvægt: 198,31 g/mol Kogepunkt: 130°C Smeltepunkt: –7 °C Flammepunkt: 165°C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00006651 CAS-nummer: 713-95-1 EINECS: 211-932-4 |
![]() |
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...