4-iodphenylboronsyre (contains varying amounts of Anhydride)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
TCIAI0830-5GEA
1810
DKK
TCIAI0830-5G
4-iodphenylboronsyre (contains varying amounts of Anhydride)
4-iodphenylboronsyre
Formel:
C₆H₆BIO₂ Molekylvægt: 247,83 g/mol Smeltepunkt: 286°C |
MDL Number:
MFCD01319014 CAS-nummer: 5122-99-6 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 108.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...