Pyridin-2,4-dicarboxylsyre Hydrat ≥98.0% (by GC, titration analysis)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
TCIAP0553-5GEA
230
DKK
TCIAP0553-5G
Pyridin-2,4-dicarboxylsyre Hydrat ≥98.0% (by GC, titration analysis)
Pyridin-2,4-dicarboxylsyre Hydrat
Formel:
C₇H₅NO₄·xH₂O Molekylvægt: 167,12 g/mol (anhy) |
MDL Number:
MFCD00006296 CAS-nummer: 499-80-9 EINECS: 207-892-2 |
![]() |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % (calcd.on anh.substance) |
Water | 4.0 to 12.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...