Dichlor(methyl)silan ≥98.0% (by GC, titration analysis)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
TCIAD0380-25GEA
222
DKK
TCIAD0380-25G
Dichlor(methyl)silan ≥98.0% (by GC, titration analysis)
Dichlor(methyl)silan
Formel:
CH₄Cl₂Si Molekylvægt: 115,03 g/mol Kogepunkt: 41°C Smeltepunkt: –92,8 °C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00000494 CAS-nummer: 75-54-7 EINECS: 200-877-1 UN nummer: 1242 ADR: 4.3,I REACH: 01-2119436669-21 |
![]() |
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...