(3-Chlorpropyl)trichlorsilan ≥97.0% (by GC)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

TCIAC0844-25GEA 386 DKK
TCIAC0844-25G
(3-Chlorpropyl)trichlorsilan ≥97.0% (by GC)
(3-Chloropropyl)trichlorosilane
Formel: ClH₂CCH₂CH₂SiCl₃
Molekylvægt: 211,98 g/mol
Kogepunkt: 178°C
Flammepunkt: 84°C
MDL Number: MFCD00013694
CAS-nummer: 2550-06-3
EINECS: 219-844-8
UN nummer: 2987
ADR: 8,II
https://dk.vwr.com/stibo/web/std.lang.all/79/84/28937984.jpg

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR