Diphenylgermaniumdichlorid ≥95.0% (by GC)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

TCIAD1824-1GEA 657 DKK
TCIAD1824-1G
Diphenylgermaniumdichlorid ≥95.0% (by GC)
Diphenylgermaniumdichlorid
Formel: C₁₂H₁₀Cl₂Ge
Molekylvægt: 297,73 g/mol
MDL Number: MFCD00013587
CAS-nummer: 1613-66-7
EINECS: 216-562-7
UN nummer: 3265
ADR: 8,II
https://dk.vwr.com/stibo/web/std.lang.all/86/09/28978609.jpg

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR