2-(Phenoxymethyl)benzoesyre ≥98.0% (by GC, titration analysis)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
TCIAP0905-25GEA
714
DKK
TCIAP0905-25G
2-(Phenoxymethyl)benzoesyre ≥98.0% (by GC, titration analysis)
2-(Phenoxymethyl)benzoesyre
Formel:
C₁₄H₁₂O₃ Molekylvægt: 228,25 g/mol |
MDL Number:
MFCD00020294 CAS-nummer: 724-98-1 EINECS: 211-967-5 |
![]() |
Specifikationer Testresultater
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 126.0 to 129.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...