Bathophenanthroline ≥99.0% (by HPLC, titration analysis), purified by sublimation

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

TCIAB2695-1GEA 1170 DKK
TCIAB2695-1G
Bathophenanthroline ≥99.0% (by HPLC, titration analysis), purified by sublimation
Bathophenanthroline
Formel: C₂₄H₁₆N₂
Molekylvægt: 332,4 g/mol
Smeltepunkt: 215…220 °C
MDL Number: MFCD00004976
CAS-nummer: 1662-01-7
EINECS: 216-767-1
https://dk.vwr.com/stibo/web/std.lang.all/42/39/28934239.jpg

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystaline
Purity (HPLC) min. 99.0 area%
Purity (Nonaqueous Titration) min. 99.0 %
Melting point 219.0 to 222.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR