4'-Chloroacetoacetanilide ≥98.0% (by titrimetric analysis)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
TCIAC0625-25GEA
312
DKK
TCIAC0625-25G
4'-Chloroacetoacetanilide ≥98.0% (by titrimetric analysis)
4'-Chloroacetoacetanilide
Formel:
C₁₀H₁₀ClNO₂ Molekylvægt: 211,6500 g/mol |
MDL Number:
MFCD00000613 CAS-nummer: 101-92-8 EINECS: 202-989-6 |
![]() |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 131.0 to 133.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...