3-Chloro-2,5-dimethylpyrazine ≥95.0% (by GC)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
TCIAC2040-5GEA
739
DKK
TCIAC2040-5G
3-Chloro-2,5-dimethylpyrazine ≥95.0% (by GC)
3-Chloro-2,5-dimethylpyrazine
Formel:
C₆H₇ClN₂ Molekylvægt: 142,5900 g/mol Kogepunkt: 114°C Flammepunkt: 83°C |
MDL Number:
MFCD00006143 CAS-nummer: 95-89-6 EINECS: 202-463-6 |
Specifikationer Testresultater
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...