5-Bromo-2-tert-butylpyrimidine ≥98.0% (by GC, total nitrogen)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

TCIAB3079-1GEA 1580 DKK
TCIAB3079-1G
5-Bromo-2-tert-butylpyrimidine ≥98.0% (by GC, total nitrogen)
5-Bromo-2-tert-butylpyrimidine
Formel: C₈H₁₁BrN₂
Kogepunkt: 90°C
Opbevaringstemperatur: Fryser
CAS-nummer: 85929-94-8
https://dk.vwr.com/stibo/web/std.lang.all/45/20/28934520.jpg

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 51.0 to 55.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR