3-Amino-5-tert-butyl-1H-pyrazole ≥98.0% (by GC, titration analysis)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

TCIAA2101-1GEA 427 DKK
TCIAA2101-1G
3-Amino-5-tert-butyl-1H-pyrazole ≥98.0% (by GC, titration analysis)
3-Amino-5-tert-butyl-1H-pyrazole
Formel: C₇H₁₃N₃
Molekylvægt: 139,20
MDL Number: MFCD00067987
CAS-nummer: 82560-12-1
EINECS: 450-360-1
https://dk.vwr.com/stibo/web/std.lang.all/02/58/28930258.jpg

Bestil nu

Specifikationer Testresultater

Appearance White to Brown powder to crystalline
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 76.0 to 80.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR