3-Ethoxysalicylaldehyde ≥97.0% (by GC)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
TCIAE0253-25GEA
173
DKK
TCIAE0253-25G
3-Ethoxysalicylaldehyde ≥97.0% (by GC)
3-Ethoxysalicylaldehyde
Formel:
C₉H₁₀O₃ Molekylvægt: 166,18 g/mol Kogepunkt: 264°C |
MDL Number:
MFCD00003323 CAS-nummer: 492-88-6 EINECS: 207-765-1 |
Specifikationer Testresultater
Appearance | Light yellow to Brown to Dark green powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 95.0 % |
Melting point | 63.0 to 67.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...