Nitrogen cylinder, emptied/unwashed
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
VWRC600733U.EMPTYEA
0
DKK
VWRC600733U.EMPTY
Nitrogen cylinder, emptied/unwashed
Nitrogen cylinder, emptied/unwashed
Formel:
N₂ |
CAS-nummer:
7727-37-9 EINECS: 231-783-9 UN nummer: 1066 ADR: 2, Merck Index: 12,06698 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...