Carbon monoxide cylinder, emptied/unwashed
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
VWRC600183M.EMPTYEA
0
DKK
VWRC600183M.EMPTY
Carbon monoxide cylinder, emptied/unwashed
Carbon monoxide cylinder, emptied/unwashed
Formel:
CO |
CAS-nummer:
630-08-0 EINECS: 211-128-3 UN nummer: 1016 ADR: 2, Merck Index: 12,01861 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...