Hydrogen chloride cylinder, emptied/unwashed

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

VWRC600543Q.EMPTYEA 0 DKK
VWRC600543Q.EMPTY
Hydrogen chloride cylinder, emptied/unwashed
Hydrogen chloride cylinder, emptied/unwashed
Formel: HCl
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011324
CAS-nummer: 7647-01-0
EINECS: 231-595-7
UN nummer: 1789
ADR: 8,II

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR