4-Chloro-2,5-dimethoxyaniline ≥98.0% (by GC, titration analysis)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
TCIAC1996-25GEA
197
DKK
TCIAC1996-25G
4-Chloro-2,5-dimethoxyaniline ≥98.0% (by GC, titration analysis)
4-Chloro-2,5-dimethoxyaniline
Formel:
C₈H₁₀ClNO₂ Molekylvægt: 187,63 g/mol |
MDL Number:
MFCD00014893 CAS-nummer: 6358-64-1 EINECS: 228-782-0 |
![]() |
Specifikationer Testresultater
Appearance | White to Gray to Red powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 118.0 to 122.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...