2-Methoxyethyl chloroacetate ≥97.0% (by GC)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

TCIAC1346-5GEA 731 DKK
TCIAC1346-5G
2-Methoxyethyl chloroacetate ≥97.0% (by GC)
2-Methoxyethyl chloroacetate
Formel: C₅H₉ClO₃
Kogepunkt: 98°C
MDL Number: MFCD00191626
CAS-nummer: 13361-36-9
https://dk.vwr.com/stibo/web/std.lang.all/85/89/28938589.jpg

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR