Cyclopropanesulphonamide ≥98.0% (by GC)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
TCIAC2628-1GEA
378
DKK
TCIAC2628-1G
TCIAC2628-5G
Cyclopropanesulphonamide ≥98.0% (by GC)
Cyclopropanesulphonamide
Formel:
C₃H₇NO₂S Molekylvægt: 121,15 g/mol |
MDL Number:
MFCD08705286 CAS-nummer: 154350-29-5 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 106.0 to 110.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...