N-(3-Fluorobenzyl)-2,4-dimethoxyaniline, 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

H57619.03EA 1800 DKK
ALFAH57619.03 ALFAH57619.MD
N-(3-Fluorobenzyl)-2,4-dimethoxyaniline, 97%
Purity: 97%
CAS No.: 1021092-55-6
Molecular Formula: C15H16FNO2
Formula Weight: 261.29
MDL No.: MFCD12551689
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR