2-Ethoxy-N-(3-fluorobenzyl)aniline, 97%
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
H57696.MDEA 988 DKK
ALFAH57696.MD ALFAH57696.03
2-Ethoxy-N-(3-fluorobenzyl)aniline, 97%
Purity: 97%
CAS No.: 1019550-62-9
Molecular Formula: C15H16FNO
Formula Weight: 245.29
MDL No.: MFCD11141664