N-(4-Fluorobenzyl)-2-iodoaniline, 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

H57172.03EA 1320 DKK
ALFAH57172.03 ALFAH57172.MD
N-(4-Fluorobenzyl)-2-iodoaniline, 97%
Purity: 97%
CAS No.: 1040308-57-3
Molecular Formula: C13H11FIN
Formula Weight: 327.14
MDL No.: MFCD11179798
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR