2,5-Dichloro-N-(2-fluorobenzyl)aniline, 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

H57565.03EA 2470 DKK
ALFAH57565.03 ALFAH57565.MD
2,5-Dichloro-N-(2-fluorobenzyl)aniline, 97%
Purity: 97%
CAS No.: 940367-81-7
Molecular Formula: C13H10Cl2FN
Formula Weight: 270.13
MDL No.: MFCD11058508
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR