2-Fluoro-N-(2-fluorobenzyl)-5-methylaniline, 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

H57667.03EA 2200 DKK
ALFAH57667.03 ALFAH57667.MD
2-Fluoro-N-(2-fluorobenzyl)-5-methylaniline, 97%
Purity: 97%
CAS No.: 1039824-40-2
Molecular Formula: C14H13F2N
Formula Weight: 233.26
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR